Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053747.2 | NP_446199.2 | 4 | 616 | ||
XM_006253551.2 | XP_006253613.1 | 4 | 664 | |||
GenBank mRNA | BC072477.1 | - | 4 | 616 | 7103662 | |
D87950.1 | - | 4 | 476 | |||
FQ222353.1 | - | 4 | 643 |