Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053722.2 | NP_446174.1 | 2, 3 | 286 | ||
XM_006243899.2 | XP_006243961.1 | 8, 9 | 948 | |||
XM_006243901.2 | XP_006243963.1 | 8, 9 | 948 | |||
XM_006243904.2 | XP_006243966.1 | 8, 9 | 948 | |||
XM_006243905.2 | XP_006243967.1 | 8, 9 | 948 | |||
XM_006243906.2 | XP_006243968.1 | 8, 9 | 948 | |||
XM_006243910.2 | XP_006243972.1 | 2, 3 | 1196 | |||
XM_006243912.2 | XP_006243974.1 | 2, 3 | 1193 | |||
XM_006243914.2 | XP_006243976.1 | 8, 9 | 1062 | |||
XM_008766606.1 | XP_008764828.1 | 8, 9 | 948 | |||
XM_008766607.1 | XP_008764829.1 | 8, 9 | 948 | |||
XM_008766608.1 | XP_008764830.1 | 8, 9 | 948 | |||
XM_008766609.1 | XP_008764831.1 | 8, 9 | 948 | |||
XM_008766610.1 | XP_008764832.1 | 8, 9 | 948 | |||
XM_008766611.1 | XP_008764833.1 | 8, 9 | 948 | |||
XM_008766612.1 | XP_008764834.1 | 8, 9 | 948 | |||
XM_008766613.1 | XP_008764835.1 | 2, 3 | 1198 | |||
XM_008766614.1 | XP_008764836.1 | 2, 3 | 1465 | |||
XM_008766615.1 | XP_008764837.1 | 8, 9 | 948 | |||
GenBank mRNA | AJ288060.1 | - | 2, 3 | 286 |