Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053765.2 | NP_446217.1 | 3 | 559 | ||
XM_006238006.2 | XP_006238068.1 | 3 | 377 | |||
XM_006238008.1 | XP_006238070.1 | 3 | 343 | |||
XM_006238009.2 | XP_006238071.1 | 3 | 720 | |||
GenBank mRNA | BC062011.1 | - | 3 | 559 | 5621384 | |
FQ224587.1 | - | 2 | 222 | |||
Y07744.1 | - | 2 | 267 |