Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053772.2 | NP_446224.1 | 4, 5 | 538 | ||
XM_006232153.2 | XP_006232215.1 | 2, 3 | 734 | |||
XM_006232155.2 | XP_006232217.1 | 3, 4 | 385 | |||
XM_006232156.2 | XP_006232218.1 | 3, 4 | 2437 | |||
XM_008760857.1 | XP_008759079.1 | 3, 4 | 862 | |||
GenBank mRNA | FM055544.1 | - | 4, 5 | 543 | ||
FQ211701.1 | - | 3, 4 | 445 | |||
L02615.1 | - | 2, 3 | 221 |