Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053657.1 | NP_446109.1 | 4, 5 | 972 | ||
GenBank mRNA | AF021935.1 | - | 4, 5 | 972 |