Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160.2 | NP_001151.1 | 26 | 4626 | ||
NM_013229.2 | NP_037361.1 | 27 | 4755 | |||
NM_181861.1 | NP_863651.1 | 27 | 4788 | |||
NM_181868.1 | NP_863658.1 | 26 | 4659 | |||
NM_181869.1 | NP_863659.1 | 8 | 2143 | |||
GenBank mRNA | AB007873.2 | - | 26 | 4178 | ||
AF013263.1 | - | 26 | 4626 | |||
BC136531.1 | - | 27 | 4359 | 9020521 | ||
BC136532.1 | - | 27 | 4359 | 9020522 | ||
BC150255.1 | - | 26 | 4207 | 9007183 |