Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053727.2 | NP_446179.2 | 2 | 1579 | ||
XM_006253667.2 | XP_006253729.1 | 2 | 2253 | |||
XM_006253668.2 | XP_006253730.1 | 2 | 1634 | |||
XM_006253669.2 | XP_006253731.1 | 2 | 2816 | |||
XM_006253670.2 | XP_006253732.1 | 2 | 1988 | |||
GenBank mRNA | AY004663.1 | - | 1 | 1415 | ||
BC072527.1 | - | 2 | 1579 | 7097417 |