Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053735.1 | NP_446187.1 | 7 | 1196 | ||
GenBank mRNA | AY029199.1 | - | 7 | 1196 |