Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Becn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034117.1 | NP_001029289.1 | 4, 5 | 464 | ||
NM_053739.2 | NP_446191.1 | 4, 5 | 486 | |||
GenBank mRNA | AY033824.1 | - | 3, 4 | 262 | ||
BC074011.1 | - | 4, 5 | 351 | 7100185 | ||
CK600131.1 | - | 4, 5 | 464 |