Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053777.1 | NP_446229.1 | 9, 10 | 2082 | ||
GenBank mRNA | AF092450.1 | - | 8, 9 | 1817 | ||
AF108959.1 | - | 8, 9 | 2020 | |||
AF109772.1 | - | 8, 9 | 1929 | |||
AF109773.1 | - | 8, 9 | 1992 | |||
AF109774.1 | - | 9, 10 | 2082 | |||
DQ377223.1 | - | 8, 9 | 1892 |