Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053787.2 | NP_446239.2 | 15 | 2120 | ||
XM_006236376.2 | XP_006236438.1 | 13 | 1696 | |||
GenBank mRNA | AB032019.1 | - | 15 | 2120 | ||
AF209196.1 | - | 2071 | ||||
AY762624.1 | - | 14 | 1808 |