Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102548514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235165.2 | XP_006235227.1 | 2, 3 | 341 |
Gene Symbol: Crnkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053797.2 | NP_446249.1 | 1 | 294 | ||
GenBank mRNA | AF245018.1 | - | 1 | 284 | ||
BC085718.1 | - | 1 | 294 | 7190212 |