Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053817.2 | NP_446269.2 | 2 | 1289 | ||
XM_008764731.1 | XP_008762953.1 | 1 | 542 | |||
GenBank mRNA | L14851.1 | - | 2 | 1289 |