Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053812.1 | NP_446264.1 | 3 | 343 | ||
XM_006256101.2 | XP_006256163.1 | 3 | 362 | |||
GenBank mRNA | AF259504.1 | - | 3 | 343 | ||
BC089784.1 | - | 3 | 304 | 7366192 |