Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053822.2 | NP_446274.2 | 1 | 135 | ||
XM_006232565.2 | XP_006232627.1 | 2 | 235 | |||
GenBank mRNA | L18891.1 | - | 1 | 141 |