Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053859.2 | NP_446311.1 | 3, 4 | 562 | ||
GenBank mRNA | EU253553.1 | - | 4, 5 | 527 | ||
FQ211811.1 | - | 575 | ||||
U07609.1 | - | 3, 4 | 565 |