Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053863.1 | NP_446315.1 | 4, 5 | 426 | ||
XM_006229422.2 | XP_006229484.1 | 6, 7 | 1451 | |||
XM_006229423.2 | XP_006229485.1 | 6, 7 | 1563 | |||
GenBank mRNA | U10279.1 | - | 4, 5 | 426 |