Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172643.1 | NP_001166114.1 | 3, 4 | 154 | ||
NM_001271934.1 | NP_001258863.1 | 4, 5 | 685 | |||
NM_001271935.1 | NP_001258864.1 | 3, 4 | 154 | |||
NM_001272050.1 | NP_001258979.1 | 3, 4 | 364 | |||
NM_004614.4 | NP_004605.4 | 3, 4 | 590 | |||
NR_073520.1 | 2, 3 | 1518 | ||||
GenBank mRNA | AF521891.1 | - | 3, 4 | 590 | ||
AK302976.1 | - | 3, 4 | 368 | |||
BC113928.1 | - | 3, 4 | 133 | |||
BC134344.1 | - | 3, 4 | 133 | |||
BC142970.1 | - | 4, 5 | 486 | 8860391 | ||
DA500158.1 | - | 4, 5 | 280 | |||
U77088.1 | - | 3, 4 | 154 | |||
U80628.1 | - | 2, 3 | 905 | |||
Y10498.3 | - | 3, 4 | 355 |