Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014550.3 | NP_055365.2 | 3 | 620 | ||
GenBank mRNA | AY028896.1 | - | 3 | 657 | ||
AY032928.1 | - | 3 | 620 | |||
BC113659.1 | - | 3 | 638 | 8322711 | ||
BC117403.1 | - | 3 | 638 | 40125954 |