Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053824.2 | NP_446276.1 | 10 | 880 | ||
XM_006235220.2 | XP_006235282.1 | 9 | 940 | |||
XM_006235221.2 | XP_006235283.1 | 9 | 981 | |||
GenBank mRNA | BC091130.1 | - | 10 | 880 | 7367296 | |
L15618.1 | - | 1497 |