Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037315.1 | NP_001032392.1 | 3 | 1073 | ||
NM_001037316.1 | NP_001032393.1 | 2 | 831 | |||
NM_053853.2 | NP_446305.1 | 1 | 1226 | |||
XM_006253001.1 | XP_006253063.1 | 3 | 897 | |||
XM_006253002.2 | XP_006253064.1 | 1 | 756 | |||
XM_006253003.1 | XP_006253065.1 | 3 | 901 | |||
XM_006253004.2 | XP_006253066.1 | 3 | 1112 | |||
GenBank mRNA | BC078765.1 | - | 2 | 832 | 7127165 | |
U01343.1 | - | 3 | 1073 | |||
U01344.1 | - | 1 | 1724 | |||
U17260.1 | - | 2 | 893 |