Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053854.1 | NP_446306.1 | 3 | 929 | ||
XM_006253007.1 | XP_006253069.1 | 2 | 2520 | |||
XM_006253008.1 | XP_006253070.1 | 1 | 615 | |||
XM_006253009.1 | XP_006253071.1 | 2 | 1707 | |||
XM_008771063.1 | XP_008769285.1 | 2 | 694 | |||
GenBank mRNA | U01348.1 | - | 3 | 929 | ||
U17261.1 | - | 1 | 624 |