Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318082.1 | NP_001305011.1 | 2 | 266 | ||
NM_001318083.1 | NP_001305012.1 | 2 | 204 | |||
NM_001318084.1 | NP_001305013.1 | 1 | 114 | |||
NM_014569.3 | NP_055384.1 | 2 | 325 | |||
NM_145102.3 | NP_659570.1 | 2 | 204 | |||
XM_017011918.1 | XP_016867407.1 | 4 | 663 | |||
XM_017011919.1 | XP_016867408.1 | 3 | 706 | |||
XM_017011920.1 | XP_016867409.1 | 1 | 871 | |||
XM_017011921.1 | XP_016867410.1 | 2 | 637 | |||
XM_017011922.1 | XP_016867411.1 | 2 | 296 | |||
XM_017011923.1 | XP_016867412.1 | 4 | 663 | |||
XM_017011924.1 | XP_016867413.1 | 2 | 325 | |||
XM_017011926.1 | XP_016867415.1 | 2 | 364 | |||
XM_017011927.1 | XP_016867416.1 | 2 | 296 | |||
GenBank mRNA | AB023232.1 | - | 2 | 266 | ||
AF170025.1 | - | 2 | 178 | |||
AK027714.1 | - | 2 | 294 | |||
BC030790.2 | - | 2 | 235 | 4827870 | ||
BX648490.1 | - | 1 | 867 | |||
CX163381.1 | - | 2 | 171 | |||
DA044307.1 | - | 2 | 313 | |||
DA572286.1 | - | 2 | 267 | |||
DA580238.1 | - | 2 | 198 | |||
DA773579.1 | - | 2 | 325 |