Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172643.1 | NP_001166114.1 | 5, 6 | 282 | ||
NM_001172644.1 | NP_001166115.1 | 4, 5 | 643 | |||
NM_001172645.1 | NP_001166116.1 | 4, 5 | 664 | |||
NM_001271934.1 | NP_001258863.1 | 6, 7 | 813 | |||
NM_001271935.1 | NP_001258864.1 | 5, 6 | 282 | |||
NM_001272050.1 | NP_001258979.1 | 5, 6 | 492 | |||
NM_004614.4 | NP_004605.4 | 5, 6 | 718 | |||
NR_073520.1 | 4, 5 | 1646 | ||||
GenBank mRNA | AF521891.1 | - | 5, 6 | 718 | ||
AK294627.1 | - | 4, 5 | 374 | |||
AK295808.1 | - | 4, 5 | 353 | |||
AK302976.1 | - | 5, 6 | 496 | |||
AK303590.1 | - | 4, 5 | 442 | |||
AK316226.1 | - | 4, 5 | 353 | |||
BC113928.1 | - | 5, 6 | 261 | |||
BC134344.1 | - | 5, 6 | 261 | |||
BC142970.1 | - | 6, 7 | 614 | 8860391 | ||
U77088.1 | - | 5, 6 | 282 | |||
U80628.1 | - | 4, 5 | 1033 | |||
Y10498.3 | - | 5, 6 | 483 |