Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053919.2 | NP_446371.1 | 3 | 579 | ||
GenBank mRNA | AF172446.1 | - | 3 | 581 | ||
FQ220766.1 | - | 3 | 579 | |||
FQ229599.1 | - | 3 | 573 |