Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053923.1 | NP_446375.1 | 17 | 2319 | ||
XM_008763348.1 | XP_008761570.1 | 17 | 2371 | |||
GenBank mRNA | AB009636.1 | - | 17 | 2319 |