Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053945.2 | NP_446397.1 | 24, 25 | 4014 | ||
NM_145881.2 | NP_665888.1 | 1, 2 | 336 | |||
XM_006241591.2 | XP_006241653.1 | 22, 23 | 4082 | |||
XM_006241592.2 | XP_006241654.1 | 21, 22 | 4016 | |||
XM_006241593.2 | XP_006241655.1 | 23, 24 | 3982 | |||
XM_006241594.2 | XP_006241656.1 | 22, 23 | 3940 | |||
XM_006241598.2 | XP_006241660.1 | 22, 23 | 6786 | |||
XM_008765437.1 | XP_008763659.1 | 25, 26 | 6284 | |||
XM_008765438.1 | XP_008763660.1 | 23, 24 | 4133 | |||
XM_008765439.1 | XP_008763661.1 | 21, 22 | 5978 | |||
XM_008765445.1 | XP_008763667.1 | 2, 3 | 1264 | |||
XM_008765446.1 | XP_008763668.1 | 2, 3 | 3197 | |||
XM_008765447.1 | XP_008763669.1 | 22, 23 | 3956 | |||
GenBank mRNA | AF199322.1 | - | 24, 25 | 4014 | ||
AF199330.1 | - | 23, 24 | 3966 | |||
AF199331.1 | - | 22, 23 | 3924 | |||
AF199335.1 | - | 1, 2 | 337 | |||
AF548738.1 | - | 22, 23 | 3437 |