Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053959.1 | NP_446411.1 | 2 | 235 | ||
XM_006254496.2 | XP_006254558.1 | 2 | 308 | |||
XM_006254497.2 | XP_006254559.1 | 2 | 308 | |||
XM_006254498.2 | XP_006254560.1 | 2 | 308 | |||
XM_006254499.2 | XP_006254561.1 | 2 | 308 | |||
XM_006254500.2 | XP_006254562.1 | 2 | 308 | |||
XM_006254501.2 | XP_006254563.1 | 2 | 308 | |||
XM_006254502.2 | XP_006254564.1 | 2 | 308 | |||
XM_006254503.2 | XP_006254565.1 | 2 | 308 | |||
XM_006254504.2 | XP_006254566.1 | 2 | 308 | |||
XM_006254505.2 | XP_006254567.1 | 2 | 308 | |||
XM_008772012.1 | XP_008770234.1 | 2 | 308 | |||
XM_008772013.1 | XP_008770235.1 | 2 | 308 | |||
XM_008772014.1 | XP_008770236.1 | 2 | 308 | |||
GenBank mRNA | BC089109.1 | - | 2 | 279 | 7311357 | |
Y13380.1 | - | 2 | 235 |