Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053977.2 | NP_446429.1 | 3 | 231 | ||
XM_008763521.1 | XP_008761743.1 | 3 | 1685 |