Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053995.3 | NP_446447.2 | 4 | 398 | ||
XM_006248478.2 | XP_006248540.1 | 4 | 454 | |||
XM_008768691.1 | XP_008766913.1 | 3 | 236 | |||
GenBank mRNA | BC085916.1 | - | 4 | 398 | 7108483 |