Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuzd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054005.1 | NP_446457.1 | 2 | 398 | ||
GenBank mRNA | AF022147.1 | - | 2 | 398 | ||
AF167170.1 | - | 2 | 164 | |||
BC090345.1 | - | 2 | 144 |