Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHCR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014762.3 | NP_055577.1 | 2 | 389 | ||
GenBank mRNA | AB073391.1 | - | 3 | 264 | ||
AF261758.1 | - | 2 | 359 | |||
AK302774.1 | - | 2 | 220 | |||
AK304421.1 | - | 2 | 338 | |||
AK304437.1 | - | 2 | 106 | |||
AK316419.1 | - | 2 | 151 | |||
AK316427.1 | - | 2 | 90 | |||
BC004375.1 | - | 2 | 295 | 3638639 | ||
BC011669.2 | - | 2 | 322 | 4130396 | ||
BP200916.1 | - | 2 | 389 | |||
D13643.3 | - | 2 | 372 |