Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS2 SIRNA Assays ›
Gene Symbol:
Rims2
Gene Name:
regulating synaptic membrane exocytosis 2
Gene Aliases:
Nim2, Rim2
Chromosome Location:
Chr.7: 78092020 - 78594162 on Build Rnor_6.0
UniGene ID:
Rn.161948
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
56114
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rims2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053945.2
NP_446397.1
25
4017
NM_145881.2
NP_665888.1
2
339
XM_006241591.2
XP_006241653.1
23
4085
XM_006241592.2
XP_006241654.1
22
4019
XM_006241593.2
XP_006241655.1
24
3985
XM_006241594.2
XP_006241656.1
23
3943
XM_006241598.2
XP_006241660.1
23
6789
XM_008765437.1
XP_008763659.1
26
6287
XM_008765438.1
XP_008763660.1
24
4136
XM_008765439.1
XP_008763661.1
22
5981
XM_008765440.1
XP_008763662.1
19, 20
3566
XM_008765441.1
XP_008763663.1
20, 21
3473
XM_008765442.1
XP_008763664.1
19, 20
3425
XM_008765443.1
XP_008763665.1
18, 19
3358
XM_008765445.1
XP_008763667.1
3
1267
XM_008765446.1
XP_008763668.1
3
3200
XM_008765447.1
XP_008763669.1
23
3959
GenBank mRNA
AF199322.1
-
25
4017
AF199323.1
-
19, 20
3408
AF199324.1
-
19, 20
3549
AF199325.1
-
20, 21
3456
AF199326.1
-
19, 20
3408
AF199327.1
-
19, 20
3408
AF199328.1
-
18, 19
3228
AF199329.1
-
18, 19
3342
AF199330.1
-
24
3969
AF199331.1
-
23
3927
AF199332.1
-
19, 20
3408
AF199335.1
-
2
340
AF548738.1
-
23
3440
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
positive regulation of gene expression
calcium ion regulated exocytosis
regulation of exocytosis
cAMP-mediated signaling
insulin secretion
regulation of membrane potential
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of inhibitory postsynaptic potential
positive regulation of excitatory postsynaptic potential
protein binding
Rab GTPase binding
protein domain specific binding
ion channel binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0