Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053992.1 | NP_446444.1 | 7 | 1830 | ||
GenBank mRNA | U41662.1 | - | 7 | 1830 |