Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054006.1 | NP_446458.1 | 3 | 352 | ||
GenBank mRNA | FQ222050.1 | - | 1 | 44 | ||
FQ223276.1 | - | 3 | 350 | |||
X52311.1 | - | 3 | 352 |
Gene Symbol: LOC100364335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751853.3 | XP_003751901.1 | 1 | 353 |