Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057131.1 | NP_476472.1 | 4, 5 | 408 | ||
XM_006246434.2 | XP_006246496.1 | 4, 5 | 409 | |||
XM_006246435.2 | XP_006246497.1 | 5, 6 | 615 | |||
XM_006246436.2 | XP_006246498.1 | 4, 5 | 400 | |||
XM_006246437.2 | XP_006246499.1 | 3, 4 | 362 | |||
XM_006246438.2 | XP_006246500.1 | 3, 4 | 283 | |||
XM_006246440.2 | XP_006246502.1 | 4, 5 | 483 | |||
GenBank mRNA | D84434.1 | - | 4, 5 | 408 | ||
FQ212452.1 | - | 4, 5 | 356 |