Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057136.1 | NP_476477.1 | 3 | 569 | ||
GenBank mRNA | AF018261.1 | - | 3 | 569 |