Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053967.2 | NP_446419.1 | 2, 3 | 1285 | ||
GenBank mRNA | BC081748.1 | - | 2, 3 | 1226 | 7134081 | |
X89999.1 | - | 2, 3 | 1289 |