Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053978.1 | NP_446430.1 | 7 | 611 | ||
XM_008770195.1 | XP_008768417.1 | 8 | 1075 | |||
XM_008770197.1 | XP_008768419.1 | 7 | 1189 | |||
GenBank mRNA | BC072512.1 | - | 8 | 830 | 7104824 | |
X78606.1 | - | 7 | 611 |