Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184796.1 | NP_001171725.1 | 2 | 540 | ||
NM_015292.2 | NP_056107.1 | 2 | 540 | |||
GenBank mRNA | AB018290.1 | - | 2 | 297 | ||
AK022790.1 | - | 2 | 491 | |||
AK225035.1 | - | 2 | 475 | |||
AK290337.1 | - | 2 | 475 | |||
BC004998.1 | - | 2 | 472 | 2958094 | ||
DB100366.1 | - | 2 | 541 | |||
DQ993200.1 | - | 2 | 422 |