Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054004.2 | NP_446456.1 | 5 | 935 | ||
GenBank mRNA | D87671.1 | - | 5 | 937 |