Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057118.2 | NP_476459.1 | 9 | 1104 | ||
XM_006242177.2 | XP_006242239.1 | 9 | 1119 | |||
XM_008765659.1 | XP_008763881.1 | 8 | 1045 | |||
GenBank mRNA | D38492.1 | - | 9 | 1108 |