Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057120.1 | NP_476461.1 | 1 | 146 | ||
XM_006248911.2 | XP_006248973.1 | 2 | 197 | |||
GenBank mRNA | D49977.1 | - | 1 | 146 |