Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr0b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057133.1 | NP_476474.1 | 1 | 562 | ||
GenBank mRNA | BC088117.1 | - | 1 | 549 | 7324177 | |
D86580.1 | - | 1 | 562 |