Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057142.1 | NP_476483.1 | 16 | 1600 | ||
GenBank mRNA | EU348625.1 | - | 3 | 215 | ||
EU348626.1 | - | 2 | 167 | |||
EU348630.1 | - | 2 | 167 | |||
EU348631.1 | - | 2 | 179 | |||
EU348632.1 | - | 2 | 164 | |||
EU348633.1 | - | 2 | 189 | |||
EU348634.1 | - | 1 | 80 | |||
EU348635.1 | - | 2 | 191 | |||
EU348636.1 | - | 2 | 143 | |||
EU348637.1 | - | 3 | 215 | |||
EU348638.1 | - | 2 | 185 | |||
EU348640.1 | - | 15 | 1523 | |||
EU348641.1 | - | 15 | 1524 | |||
EU348643.1 | - | 16 | 1600 | |||
EU348644.1 | - | 16 | 1600 | |||
U66707.1 | - | 16 | 1600 |