Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270626.1 | NP_001257555.1 | 3 | 817 | ||
NM_001270627.1 | NP_001257556.1 | 3 | 817 | |||
NM_001270628.1 | NP_001257557.1 | 3 | 817 | |||
NM_057190.2 | NP_476538.2 | 3 | 817 | |||
XM_006233554.2 | XP_006233616.1 | 3 | 1120 | |||
XM_008761575.1 | XP_008759797.1 | 3 | 1120 | |||
XM_008761576.1 | XP_008759798.1 | 3 | 1120 | |||
GenBank mRNA | AJ293697.1 | - | 3 | 750 | ||
AJ293698.1 | - | 3 | 750 | |||
AJ293699.1 | - | 3 | 750 | |||
AJ534640.1 | - | 3 | 749 | |||
AJ534642.1 | - | 3 | 749 | |||
BC087719.1 | - | 3 | 778 | 7318793 |