Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3MBTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015478.6 | NP_056293.4 | 1 | 283 | ||
NM_032107.4 | NP_115479.4 | 4 | 454 | |||
GenBank mRNA | AK299199.1 | - | 4 | 424 | ||
AK308334.1 | - | 1 | 250 | |||
AK310345.1 | - | 1 | 283 | |||
BC039820.1 | - | 1 | 198 | 5199689 | ||
DA204976.1 | - | 3 | 373 |