Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Decr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057197.2 | NP_476545.2 | 2 | 228 | ||
GenBank mRNA | BC059120.1 | - | 2 | 216 | 6920489 | |
CO385941.1 | - | 2 | 213 | |||
D00569.1 | - | 2 | 201 | |||
FQ209689.1 | - | 2 | 217 | |||
FQ210607.1 | - | 2 | 209 | |||
FQ210721.1 | - | 2 | 211 | |||
FQ210830.1 | - | 2 | 198 | |||
FQ212507.1 | - | 2 | 206 | |||
FQ218387.1 | - | 2 | 206 | |||
FQ219629.1 | - | 2 | 209 | |||
FQ219666.1 | - | 2 | 206 | |||
FQ220227.1 | - | 2 | 209 |