Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080397.1 | NP_536322.1 | 3 | 382 | ||
XM_006244730.2 | XP_006244792.1 | 4 | 531 | |||
XM_006244732.2 | XP_006244794.1 | 3 | 189 | |||
XM_008766985.1 | XP_008765207.1 | 3 | 313 | |||
XM_008766987.1 | XP_008765209.1 | 3 | 283 | |||
GenBank mRNA | AF022729.1 | - | 3 | 382 |