Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080780.2 | NP_542958.2 | 2 | 360 | ||
XM_006246566.2 | XP_006246628.1 | 2 | 440 | |||
GenBank mRNA | X92069.1 | - | 2 | 360 | ||
X97328.1 | - | 2 | 226 |